Tisk…

N-Hydroxysuccinimide ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: 1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu

A10312.14 A10312.22 A10312.36
A10312.14EA 1210 CZK
A10312.14 A10312.22 A10312.36
N-Hydroxysuccinimide ≥98%
N-Hydroxysuccinimide

Purity: 98+% CAS No.: 6066-82-6 Molecular Formula: C4H5NO3 Formula Weight: 115.09 Melting Point: 94-99° Beilstein Reference: 113913

Vzorec: C₄H₅NO₃
MW: 115.09
Bod varu: 363
Bod tání: 94…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00005516
CAS číslo: 6066-82-6
EINECS: 228-001-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR