Tisk…

Šťavelan diamonný monohydrát ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Ammonium oxalate monohydrate, Ammonium oxalate-1-hydrate, Oxalic acid diammonium salt monohydrate, Oxalic acid ammonium salt monohydrate, Ammonium ethanedioate monohydrate, Ammonium Oxalate, Monohydrate

A10263.0E A10263.22 A10263.36
A10263.0EEA 6800 CZK
A10263.0E A10263.22 A10263.36
Šťavelan diamonný monohydrát ≥98%
Šťavelan diamonný monohydrát

Purity: 98% CAS No.: 6009-70-7 Molecular Formula: C2H8N2O4.H2O Formula Weight: 142.11 (124.10anhy) Physical Form: Crystalline solid Density: 1.50 Appearance: White Melting Point: 70° dec. Beilstein Reference: 3759830 Merck Reference: 14,537 UN No.: 3288 MDL No.: MFCD00149694

Vzorec: (NH₄)₂C₂O₄·H₂O
Bod varu: 70 °C (1013 hPa)
Hustota: 1,50
Storage Temperature: Ambient
MDL Number: MFCD00149694
CAS číslo: 6009-70-7
EINECS: 238-135-4
UN: 3288
ADR: 6.1,III
Merck Index: 14,537

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR