Tisk…

4-(Bromomethyl)benzenesulphonyl chloride 95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A10120.06 A10120.14
A10120.06EA 2980 CZK
A10120.06 A10120.14
4-(Bromomethyl)benzenesulphonyl chloride 95%
4-(Bromomethyl)benzenesulphonyl chloride

Purity: 95% CAS No.: 66176-39-4 Molecular Formula: C7H6BrClO2S Formula Weight: 269.55 Melting Point: 72-75° Beilstein Reference: 2364328 UN No.: 3261 MDL No.: MFCD00156129

Vzorec: C₇H₆BrClO₂S
Bod tání: 72…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00156129
CAS číslo: 66176-39-4
UN: 3261
ADR: 8,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR