Tisk…

Pyridoxal hydrochloride

Dodavatel: PanReac AppliChem

Synonyms: 3-Hydroxy-5-(hydroxymethyl)-2-methylisonicotinaldehyde hydrochloride, Pyridoxal HCl

A0960.0025
A0960.0025EA 10860 CZK
A0960.0025
Pyridoxal hydrochloride
Pyridoxal hydrochloride
Vzorec: C₈H₁₀ClNO₃
MDL Number: MFCD00012809
CAS číslo: 65-22-5
EINECS: 200-602-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR