Tisk…
VWR®, Flash point reference standard, Pensky Martens
Dodavatel: VWR Chemicals
99155.001EA 7750 CZK
99155.001 99156.001 99157.001 99158.001 99159.001 99160.001
VWR®, Flash point reference standard, Pensky Martens
Standardy Standardy pro ropný průmysl
PMCC flash point standards are tested and certified in accordance with ASTM D93 'Standard Test Method for Flash Point by Pensky Martens Closed Cup Tester - Procedure A'
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • One bottle per test ensures zero contamination / high-end volatile loss
  • Fully traceable to international standards
  • Supplied with MSDS
  • Tamper-evident security packaging
  • 12 month shelf life
Procedure A is applicable to distillate fuels - diesels, bio-diesel blends, kerosene, heating oil, turbine fuels and lubricating oils. These UKAS certified standards are specifically designed to be used for regular instrument performance checks. Packed uniquely in 3×80 ml 'single shot' bottles ensuring the highest level of sample integrity. An economical, time saving addition to any laboratory concerned with the generation of valid analytical test data. Typically used in, but not limited to, the analysis of petroleum products with flash points above 40 °C and below 360 °C.
Certifikace: UKAS ISO 17025 and ISO 17034.
Upozornení: The flash point given in the ordering table below is the nominal flash point - actual batch values are given in the certification documents supplied with the product and also on the label.