Tisk…

2-Propanol 99,8, BAKER ANALYZED® LC-MS, J.T.Baker®

Dodavatel: Avantor
Nebezpečí

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

9827-02EA 4420 CZK
9827-02
2-Propanol 99,8, BAKER ANALYZED® LC-MS, J.T.Baker®
2-Propanol
Vzorec: (CH₃)₂CHOH
MW: 60.1
Bod varu: 82
Bod tání: –89 °C
Hustota: 0.786 g/cm³ (20 °C)
Bod vzplanutí: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS číslo: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

For Laboratory,Research,or Manufacturing Use = 0.01
UV Absorbance - 260 - 400 nm ≤0.01
UV Absorbance - 230 nm ≤0.15
UV Absorbance - 220 nm ≤0.30
Assay (CH₃CHOH CH₃) ≥99.8 %
Residue after Evaporation ≤1.0 ppm
Water (H₂O) ≤0.06 %
LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine) ≤50 ppb
Trace Impurities - Aluminum (Al) ≤50.0 ppb
Trace Impurities - * Calcium (Ca) ≤50.0 ppb
Trace Impurities - Iron (Fe) ≤50.0 ppb
Trace Impurities - Magnesium (Mg) ≤50.0 ppb
Trace Impurities - Potassium (K) ≤50 ppb
Trace Impurities - * Sodium (Na) ≤50.0 ppb
* May change over time due to extraction from glass container.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR