Tisk…

p-Xylen ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™

Dodavatel: Honeywell Chemicals
Nebezpečí

Synonyms: 1,4-Dimethylbenzene

95682-2.5L
95682-2.5LEA 7600 CZK
95682-2.5L
p-Xylen ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
p-Xylen

Beilstein Number: 1901563
RTECS Number: ZE2625000
UNSPSC Code: 12190000

Vzorec: C₆H₄(CH₃)₂
Bod varu: 137…139 °C (1,013 hPa)
Bod tání: 12 °C
Hustota: 0,86 g/cm³ (20 °C)
Bod vzplanutí: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00008556
CAS číslo: 106-42-3
EINECS: 203-396-5
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8b04563a5941cefd103ebd3048bcf8486f2bdac&vid=e5db83ad1f0cb6a583610c58cea291dcbe1be1f3&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Identity (IR) complying
Assay (GC) Min. 99.0 %
Refractive index (n 20/D) 1.494 - 1.496
Non-volatile matter Max. 0.005 %
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
o-Xylene (GC) Max. 0.5 %
m-Xylole (GC) Max. 0.5 %
Sum of the isomers (GC) Max. 0.5 %
Benzene (GC) Max. 1.0 %
Ethyl benzene (GC) Max. 1.0 %
Toluene (GC) Max. 0.5 %
Refractive Index n20/D 1.495


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR