Tisk…

Kyselina chlorovodíková 36,5 - 38,0%, BAKER ANALYZED® ACS, J.T.Baker®

Dodavatel: Avantor
Nebezpečí


Management of Change (MOC) category = R

9535-03EA 2000 CZK
9535-03 9535-33 9535-15 9535-01 9535-00 9535-02 9535-07 9535-08 9535-09
Kyselina chlorovodíková 36,5 - 38,0%, BAKER ANALYZED® ACS, J.T.Baker®
Kyselina chlorovodíková
Vzorec: HCl
Bod varu: 110 °C (1013 hPa)
Bod tání: –35 °C
Hustota: 1,18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS číslo: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Objednat

Výsledky specifikačního testu

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Store below 25°C Passes Test
Appearance Passes Test
Assay (as HCl) (by acid-base titrn) 36.5 - 38.0 %
Color (APHA) ≤ 10
Extractable Organic Substances ≤ 5 ppm
Free Chlorine (as Cl₂) ≤ 1 ppm
Residue after Ignition ≤ 3 ppm
Specific Gravity at 60°/60°F 1.185 - 1.192
Bromide (Br) ≤ 0.005 %
Phosphate (PO₄) ≤ 1 ppm
Sulfate (SO₄) ≤ 0.5 ppm
Sulfite (SO₃) ≤ 0.8 ppm
Ammonium (NH₄) ≤ 3 ppm
Trace Impurities - Aluminum (Al) ≤ 100.0 ppb
Arsenic and Antimony (as As) ≤ 5.0 ppb
Trace Impurities - Boron (B) ≤ 50 ppb
Trace Impurities - Calcium (Ca) ≤ 200.0 ppb
Trace Impurities - Chromium (Cr) ≤ 100.0 ppb
Trace Impurities - Copper (Cu) ≤ 100.0 ppb
Trace Impurities - Gold (Au) ≤ 100.0 ppb
Heavy Metals (as Pb) ≤ 100 ppb
Trace Impurities - Iron (Fe) ≤ 100.0 ppb
Trace Impurities - Lead (Pb) ≤ 50 ppb
Trace Impurities - Magnesium (Mg) ≤ 300.0 ppb
Trace Impurities - Manganese (Mn) ≤ 300.0 ppb
Trace Impurities - Mercury (Hg) ≤ 5.0 ppb
Trace Impurities - Nickel (Ni) ≤ 100.0 ppb
Trace Impurities - Potassium (K) ≤ 300.0 ppb
Trace Impurities - Sodium (Na) ≤ 300 ppb
Trace Impurities - Tin (Sn) ≤ 300.0 ppb
Trace Impurities - Titanium (Ti) ≤ 300.0 ppb
Trace Impurities - Zinc (Zn) ≤ 100.0 ppb
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR