Tisk…

Tetrahydrofuran, low water content ≥99.8% (by GC, corrected for water content) contains no preservatives for HPLC, J.T.Baker®

Dodavatel: Avantor
Nebezpečí

Synonyms: Butylene oxide, THF, Diethylene oxide

9439-12EA 0 CZK
9439-12
Tetrahydrofuran, low water content ≥99.8% (by GC, corrected for water content) contains no preservatives for HPLC, J.T.Baker®
Tetrahydrofuran

For use in Liquid Chromatography, Organic and Biosynthesis.

Vzorec: C₄H₈O
Bod varu: 66 °C (1013 hPa)
Hustota: 0,888 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS číslo: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

For Laboratory,Research,or Manufacturing Use Passes Test
Appearance (Clear, colorless liquid) Passes Test
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-320 nm ≤0.005
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.030
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm ≤0.15
Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm ≤0.40
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤212
Assay (by GC, corrected for water) ≥99.8 %
Color (APHA) ≤10
Peroxide (as H₂O₂) (at time of fill) ≤10 ppm
Residue after Evaporation ≤2.0 ppm
Water (by KF, coulometric) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR