Tisk…
Organic reference standard, Butyraldehyde, contains ~0.1% 2,6-di-tert-butyl-4-methylphenol and ~1% water as stabiliser, analytical standard, Supelco®
Dodavatel: Merck
94194-1ML
94194-1MLEA 1850 CZK
94194-1ML
Organic reference standard, Butyraldehyde, contains ~0.1% 2,6-di-tert-butyl-4-methylphenol and ~1% water as stabiliser, analytical standard, Supelco®
Standardy Standardy organické
Butyraldehyde (BA) belongs to the class of volatile organic compounds (VOCs).
Analytical standard reference:
  • Air samples by pentafluorophenyl hydrazine derivatisation followed by analysis using gas chromatography (GC) equipped with flame ionisation detector (FID)
  • Ambient air by GC coupled to mass spectrometry (MS)
  • Saliva samples by magnetic-stirring-assisted micro-solid-phase extraction (μ-SPE) and headspace GC combined with ion-mobility spectrometry (IMS)
Also used as analytical Environmental samples by 2,4-dinitrophenylhydrazine derivatisation followed by analysis using rapid resolution liquid chromatography-ultraviolet diode array detector (RRLC-UV-DAD) and RRLC coupled to ion-trap mass spectrometer (IT-MS) operating under the atmospheric pressure chemical ionisation (APCI) mode.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0721f9dff2a0f9ab1be6dbcf6c11ffd8bf86449e&vid=0721f9dff2a0f9ab1be6dbcf6c11ffd8bf86449e&prid=web&clid=SAPDAM
Objednat
Produktová specifikace
Popis Butyraldehyde
Environmentally Preferable
Čí. CAS 77220-86-1
Vzorec C₅H₁₀O₃ ·xNa⁺ ·yH₂O
Skladovací teplota 2...8 °C