Tisk…

Biocytin ≥95% (by TLC)

Dodavatel: Biotium

Synonyms: ε-Biotinoyl-L-lysine, H-Lys(biotinyl)-OH

90055
90055.EA 4680 CZK
90055.
Biocytin ≥95% (by TLC)
Biocytin

Biocytin (ε-Biotinoyl-L-lysine) is a cellular tracer that can be introduced into cells by microinjection.


  • White solid soluble in water or DMSO


Biocytin is the natural substrate of biotinidase and can be used to measure the biotinidase activity and diagnose biotinidase deficiency in scientific research as a histological stain for nerve cells.

Vzorec: C₁₆H₂₈N₄O₄S
MW: 372.484 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077319
CAS číslo: 576-19-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR