Tisk…

5(6)-TAMRA free acid ≥97% (by HPLC) amine-reactive fluorescent dye

Dodavatel: Biotium

90027
90027.EA 5980 CZK
90027.
5(6)-TAMRA free acid ≥97% (by HPLC) amine-reactive fluorescent dye
5(6)-TAMRA

Mixed isomers of carboxytetramethylrhodamine (TAMRA) free acid. The dye can be coupled to primary or secondary amines via standard peptide chemistry.


  • λEx/λEm (MeOH) = 540/565 nm
  • Dark red solid soluble in DMSO, DMF, MeOH, or pH >6.5 buffer
  • Store at -20°C and protect from light

Vzorec: C₂₅H₂₂N₂O₅
Storage Temperature: Freezer
MDL Number: MFCD16661194
CAS číslo: 98181-63-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecc9f1b2f43d5bca81fa2cd0e805d6a054386bd5&vid=ecc9f1b2f43d5bca81fa2cd0e805d6a054386bd5&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR