Tisk…

Silikagel ≥99.5%, large pore

Dodavatel: Thermo Fisher Scientific
Varování

89385.36 89385.A3
89385.36EA 3680 CZK
89385.36 89385.A3
Silikagel ≥99.5%, large pore
Silikagel

Purity: 99.5% CAS No.: 63231-67-4 Physical Form: 56 Micron, S.A. 500m2/g, P.V. 0.75cc/g Appearance: White MDL No.: MFCD00011232

Vzorec: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS číslo: 63231-67-4
EINECS: 231-545-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR