Tisk…

Hydrid titanatý ≥99% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

89183.06 89183.18 89183.30
89183.06EA 1170 CZK
89183.06 89183.18 89183.30
Hydrid titanatý ≥99% (metals basis)
Hydrid titanatý

Purity: 99% CAS No.: 7704-98-5 Molecular Formula: H2Ti Formula Weight: 49.92 Physical Form: 1-3 Micron Powder Density: 3.9 Appearance: Dark grey Odour: Odourless Melting Point: 450° dec. Merck Reference: 14,9473 UN No.: 1871 MDL No.: MFCD00151301


For preparation of titanium compounds and coatings, in formation of glass and ceramic seals, as a catalyst, for thermal storage, as a source for pure H2

Vzorec: H₂Ti
Hustota: 3,9
Storage Temperature: Ambient
MDL Number: MFCD00151301
CAS číslo: 7704-98-5
EINECS: 231-726-8
UN: 1871
ADR: 4.1,II
Merck Index: 14,9473

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR