Tisk…

Strontium chromate ≥95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

89026.30 89026.A1
89026.30EA 2790 CZK
89026.30 89026.A1
Strontium chromate ≥95%
Strontium chromate

CAS No.: 7789-06-2 Molecular Formula: SrCrO4 Formula Weight: 203.61 Physical Form: Powder Density: 3.895 Odour: Odourless Merck Reference: 14,8841 UN No.: 3087 MDL No.: MFCD00049549

Vzorec: CrO₄Sr
Hustota: 3,895
Storage Temperature: Ambient
MDL Number: MFCD00049549
CAS číslo: 7789-06-2
EINECS: 232-142-6
UN: 3087
ADR: 5.1,III
Merck Index: 14,8841

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR