Tisk…

Silikagel ≥99.5% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

88316.30 88316.A1
88316.30EA 3640 CZK
88316.30 88316.A1
Silikagel ≥99.5% (metals basis)
Silicon dioxide

Silicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.


Purity: 99.5% CAS No.: 14808-60-7 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: -325 Mesh Powder Density: 2.2-2.6 Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232

Vzorec: SiO₂
Bod varu: 2230 °C
Bod tání: 1710 °C
Hustota: 2,2…2.6
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS číslo: 14808-60-7
EINECS: 238-878-4
Merck Index: 14,8493

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR