Tisk…

Voda Reag. Ph. Eur. 1095507 Particle_free

Dodavatel: VWR Chemicals

Synonyms: Hydrogen oxide

87962.290EA 1068 CZK
87962.290
Voda Reag. Ph. Eur. 1095507 Particle_free
Voda

Informace pro objednávání: Supplied with certificate of analysis.

Vzorec: H₂O
MW: 18.02
Bod varu: 100
Bod tání: 0
Hustota: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS číslo: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR