Tisk…

Kyselina šťavelová 0,5% sulphuric acid_50 Reag. Ph. Eur. 1061401

Dodavatel: VWR Chemicals
Nebezpečí

Synonyms: Ethanedioic acid

87883.290EA 4290 CZK
87883.290
Kyselina šťavelová 0,5% sulphuric acid_50 Reag. Ph. Eur. 1061401
Kyselina šťavelová

Informace pro objednávání: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Vzorec: C₂H₂O₄
MW: 90.04
Bod varu: 365
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS číslo: 144-62-7
EINECS: 205-634-3
UN: 2796
ADR: 8 II
Merck Index: 13,06980

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR