Tisk…

Destaining solution Reag. Ph. Eur. 1012202

Dodavatel: VWR Chemicals

87788.290EA 4290 CZK
87788.290
Destaining solution Reag. Ph. Eur. 1012202
Destaining solution

The reagent is specially intended for use in testing Ph Eur and monographs. This solution was produced according 4.1.1 of Ph Eur specifications using Ph Eur reagent quality components and water meeting the Ph Eur monograph requirement for Purified Water.


Content: 1 volume of glacial acetic acid R, 4 volumes of methanol R and 5 volumes of water R


Informace pro objednávání: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Storage Temperature: Ambient
UN: 2921
ADR: 8 (4.1) II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR