Tisk…

Alizarin red S (sodium salt) solution_SC Reag. Ph. Eur. 1002601

Dodavatel: VWR Chemicals

Synonyms: C.I. 58005

87763.180EA 1830 CZK
87763.180
Alizarin red S (sodium salt) solution_SC Reag. Ph. Eur. 1002601
Alizarin red S (sodium salt)

Informace pro objednávání: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Vzorec: C₁₄H₇NaO₇S
MW: 342.26 g/mol
MDL Number: MFCD00013049
CAS číslo: 130-22-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR