Tisk…

Zinc iodine chloride in aqueous solution Reag. Ph. Eur. 1096602

Dodavatel: VWR Chemicals
Nebezpečí

87726.180EA 6620 CZK
87726.180
Zinc iodine chloride in aqueous solution Reag. Ph. Eur. 1096602
Zinc iodine chloride in aqueous solution

Informace pro objednávání: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Storage Temperature: Ambient
UN: 2920
ADR: 8 (3),II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR