Tisk…

Tetrabutylammonium tetrafluoroborate, Supelco®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ100

86896-25G
86896-25GEA 13140 CZK
86896-25G
Tetrabutylammonium tetrafluoroborate, Supelco®
Tetrabutylammonium tetrafluoroborate
Vzorec: C₁₆H₃₆BF₄N
Bod tání: 159…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00011634
CAS číslo: 429-42-5
EINECS: 207-058-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=baaa7ae03a380912632ea5df8e9df71e7fb29f9d&vid=76bd23963b9e29cb9ba7ed1fc76c2844eed3d8f6&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR