Tisk…

Thymolphthalein, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. Indicator

Dodavatel: VWR Chemicals

Synonyms: 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one

86157.050EA 1056 CZK
86157.050 86157.150 86157.180
Thymolphthalein, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. Indicator
Thymolphthalein
Vzorec: C₂₈H₃₀O₄
MW: 430.54
Bod tání: 253
Storage Temperature: Ambient
MDL Number: MFCD00005909
CAS číslo: 125-20-2
Merck Index: 12,09542

Objednat

Výsledky specifikačního testu

Identity (UV-VIS Spectroscopy) Passes Test
Appearance of Solution Passes Test
pH Range (Colourless to Blue) 8.8 - 10.5
Absorption Maximum λ (pH 10,5) 592 - 596
Absorptivity (A1%/1 cm; λ max , 0.01 g/l, pH 10.5, on Dried Sample) 800 - 900
Loss on Drying (110 °C) ≤1.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR