Tisk…

Ethanol eurodenatured ≥96%, TechniSolv® (1:1:1)

Dodavatel: VWR Chemicals

85824.360EA 2130 CZK
85824.360 85824.440
Ethanol eurodenatured ≥96%, TechniSolv® (1:1:1)
Ethanol Eurodenatured
Vzorec: H₃CCH₂OH
CAS číslo: 64-17-5
UN: 1170
ADR: 3,II

Objednat

Výsledky specifikačního testu

Assay (V/V) min. 93.8%
Denatonium benzoate (ppm) 11.8 - 13.8 ppm
Methyl ethyl ketone (V/V) 0.93 - 1.09 %
2-Propanol (V/V) 0.93 - 1.09%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR