Tisk…

Uhličitan draselný aqueous solution USP Test Solution (TS)

Dodavatel: VWR Chemicals

Synonyms: Potash

85365.180EA 899 CZK
85365.180 85365.260
Uhličitan draselný aqueous solution USP Test Solution (TS)
Uhličitan draselný

Ready to use test solution (TS).


  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available

Vzorec: K₂CO₃
MW: 138.21
Bod tání: 891
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS číslo: 584-08-7
EINECS: 0
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR