Tisk…

Potassium bromate 0,100 N in aqueous solution 1 N in aqueous solution USP Test Solution (TS) 34

Dodavatel: VWR Chemicals

85334.180EA 4540 CZK
85334.180 85334.260
Potassium bromate 0,100 N in aqueous solution 1 N in aqueous solution USP Test Solution (TS) 34
broman draselný

Ready to use test solutions (TS).


  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available

Vzorec: KBrO₃
Bod tání: -7.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00011359
CAS číslo: 7758-01-2
EINECS: 0
ADR: 8,I
Merck Index: 13,01378

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR