Tisk…

Kyselina sírová 93% 93, NORMATOM® Trace_metal

Dodavatel: VWR Chemicals
Nebezpečí

83875.270EA 6590 CZK
83875.270 83875.330 83875.290
Kyselina sírová 93% 93, NORMATOM® Trace_metal
Kyselina sírová
Vzorec: H₂SO₄
MW: 98.08
Bod varu: 330
Bod tání: 10.38
Hustota: 1.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS číslo: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Objednat

Výsledky specifikačního testu

Assay 93 - 98 %
Colouration Max. 10 APHA
Cl (Chloride)
Total P (Phosphorus)
NO3 (Nitrate)
Substances reducing KMnO4
Ag (Silver) Max. 1 ppb
Al (Aluminium) Max. 1 ppb
As (Arsenic)
Au (Gold)
Ba (Barium) Max. 1 ppb
Be (Beryllium)
Bi (Bismuth)
Ca (Calcium) Max. 1 ppb
Cd (Cadmium)
Ce (Cerium)
Co (Cobalt)
Cr (Chromium)
Cs (Cesium)
Dy (Dysprosium)
Er (Erbium)
Eu (Europium)
Ga (Gallium)
Gd (Gadolinium)
Ge (Germanium) Max. 1 ppb
Hf (Hafnium)
Hg (Mercury)
Ho (Holmium)
In (Indium)
La (Lanthanum)
Li (Lithium)
Lu (Lutetium)
Mg (Magnesium) Max. 1 ppb
Mn (Manganese)
Mo (Molybdenum)
Na (Sodium) Max. 1 ppb
Nb (Niobium)
Nd (Neodymium)
Ni (Nickel)
Pr (Praseodymium)
Rb (Rubidium)
Rh (Rhodium)
Sb (Antimony) Max. 1 ppb
Sc (Scandium)
Se (Selenium) Max. 10 ppb
Sm (Samarium)
Sn (Tin) Max. 1 ppb
Sr (Strontium)
Tb (Terbium)
Te (Tellurium)
Th (Torio) Max. 0,100 ppb
Ti (Titanium) Max. 1 ppb
Tl (Thallium)
Tm (Thulium)
U (Uranium)
V (Vanadium)
Y (Yttrium)
Yb (Ytterbium)
Zn (Zinc) Max. 1 ppb
Zr (Zirconium)

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR