Tisk…

Octan rtuťnatý ≥99.0% (by precipitation titration), Puriss. p.a. ACS, Fluka™

Dodavatel: Honeywell Chemicals
Nebezpečí

83352-250G 83352-50G
83352-250GEA 6490 CZK
83352-250G 83352-50G
Octan rtuťnatý ≥99.0% (by precipitation titration), Puriss. p.a. ACS, Fluka™
Octan rtuťnatý
Vzorec: (H₃CCOO)₂Hg
Hustota: 3,27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012165
CAS číslo: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1,II
Merck Index: 13,05898

Objednat

Výsledky specifikačního testu

Assay 99.0 - 101.0 %
Insoluble in acetic acid Max. 0.01 %
Residue on ignition a. reducing with HCOOH (SO₄)* Max. 0.02 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 10 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Mercury (Hg⁺) Max. 0.4 %
Other heavy metals (as Pb) Max. 0.002 %
Chloride (Cl) Max. 0.005 %
Nitrate (NO₃) Max. 50 ppm
Sulfate (SO₄) Max. 50 ppm

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR