Tisk…
Cryo-Babies® and Cryo-Tags® cryo label rolls
Dodavatel: DIVERSIFIED BIOTECH
CRY-1000 CRY-2000 CRY-0500
817-2131EA 2790 CZK
817-2131 DIVECRY-2000 817-0105
Cryo-Babies® and Cryo-Tags® cryo label rolls
Štítky
These Cryo-Babies® and Cryo-Tags® PVC cryo label rolls are handwritable and available in three sizes.
  • Temperature resistant from −196 to +150 °C
  • Chemically inert, resist most organic solvents and caustic agents
  • Adhere to most plastics, glass and metals without cracking, peeling or degrading
  • Easy to write on with most lab markers
Cryo-Babies® and Cryo-Tags® withstand freezer temperatures down to −196 °C (liquid and vapor phase nitrogen), boiling water baths (100 °C), autoclaving and moderate oven temperatures (150 °C).
The dispensing box assists inMore Product Information