Tisk…

Diethylether Stabilised, BAKER, J.T.Baker®

Dodavatel: Avantor
Nebezpečí

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

Stabilization: BHT

8106.1000EA 1630 CZK
8106.1000 8106.5000 8106.9025 8106.9200
Diethylether Stabilised, BAKER, J.T.Baker®
Diethylether
Vzorec: (CH₃CH₂)₂O
MW: 74.12
Bod varu: 35
Bod tání: –123 °C
Hustota: 0.7135 g/cm³ (20 °C)
Bod vzplanutí: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS číslo: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3 I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Meets Reagent Specifications for testing Ph. Eur. monographs.
Acidity passes test
Aldehydes passes test
Distillation Range 34.0 - 35.0 °C
Non-volatile Matter max. 20 mg/l
Peroxide passes test
Relative Density 0.714 - 0.716
Substances with a Foreign Odour passes test
Water (H2O) max. 2 g/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR