Tisk…

n-Hexan, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Dodavatel: Avantor
Nebezpečí

Synonyms: Hexyl hydride, Hexanen, Hexane

8044.1000EA 2170 CZK
8044.1000 8044.2500 8044.9025 8044.5000 8044.9200
n-Hexan, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
n-Hexan
Vzorec: H₃C(CH₂)₄CH₃
MW: 86.18
Bod varu: 69
Bod tání: –94.3 °C
Hustota: 0.659 g/cm³ (20 °C)
Bod vzplanutí: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS číslo: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Assay (by GC) min. 95 %
Aromatic Compounds (as C6H6) max. 0.02 %
Color (APHA) max. 10
Density (g/ml) at 25°C max. 0.657
Residue after Evaporation max. 0.001 %
Sulfur Compounds (as S) max. 0.005 %
Thiophene passes test
Water Soluble Titrable Acid (meq/g) max. 0.0003
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR