Tisk…

Ethylacetát, BAKER ANALYZED® ACS, J.T.Baker®

Dodavatel: Avantor
Nebezpečí

Synonyms: Acetic acid ethyl ester

8037.1000EA 1590 CZK
8037.1000 8037.2500 8037.9025 8037.5000
Ethylacetát, BAKER ANALYZED® ACS, J.T.Baker®
Ethylacetát
Vzorec: CH₃COOC₂H₅
MW: 88.11
Bod varu: 77.1
Bod tání: –83 °C
Hustota: 0.902
Bod vzplanutí: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS číslo: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Exceeds ACS Specifications
Assay (by GC) min. 99.5 %
Color (APHA) max. 10
Residue after Evaporation max. 0.003 %
Substances Darkened by H2SO4 passes test
Titrable Acid (meq/g) max. 0.0009
Water (H2O) max. 0.2 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR