Tisk…

Aceton, BAKER ANALYZED® ACS, J.T.Baker®

Dodavatel: Avantor
Nebezpečí

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

8002.2500EA 2710 CZK
8002.2500 8002.9025 8002.1000 8002.5000 8002.9200 8002.2500PE
Aceton, BAKER ANALYZED® ACS, J.T.Baker®
Aceton
Vzorec: CH₃COCH₃
MW: 58.08
Bod varu: 56.2
Bod tání: –95.4 °C
Hustota: 0.792 g/cm³ (20 °C)
Bod vzplanutí: <–20 °C (ot. pohárek)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS číslo: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Exceeds ACS Specifications
Assay (by GC) min. 99.5 %
Aldehyde (as HCHO) max. 0.002 %
Color (APHA) max. 10
Diisopropylether((CH3)2CHOCH(CH3)2) max. 0.05 %
Isopropyl Alcohol max. 0.05 %
Methanol (CH3OH) max. 0.05 %
Residue after Evaporation max. 5 ppm
Solubility in Water passes test
Substances Reducing KMnO4 passes test
Titrable Acid (meq/g) max. 0.0003
Titrable Base (meq/g) max. 0.0006
Water (H2O) max. 0.5 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1
at 330 nm max. 1.00
at 340 nm max. 0.10
at 350 nm max. 0.02
at 400 nm max. 0.01

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR