Tisk…
Anti-CD11b Rat Antibody (redFluor™ 710) [clone: M1/70]
Dodavatel: Tonbo Biosciences
80-0112-U025 80-0112-U100
80-0112-U025EA 2380 CZK
80-0112-U025 80-0112-U100
Anti-CD11b Rat Antibody (redFluor™ 710) [clone: M1/70]
Protilátky
The M1/70 antibody reacts with human and mouse CD11b, also known as integrin alpha M. This 165-170 kDa cell surface glycoprotein is part of a family of integrin receptors that mediate adhesion between cells (cell-cell) and components of the extracellular matrix, e.g. fibrinogen (cell-matrix). In addition, integrins are active signaling receptors which recruit leukocytes to inflammatory sites and promote cell activation. Complete, functional integrin receptors consist of distinct combinations of integrin chains which are differentially expressed. Integrin alpha M (CD11b) assembles withMore Product Information
Objednat
Produktová specifikace
Symbol antigenu ITGAM
Název antigenu Integrin Alpha M
Environmentally Preferable
Synonyma antigenu CD11B|integrin subunit alpha M|MAC-1|MO1A|MAC1A|SLEB6|CR3A
Druh protilátky Primární
Spojení redFluor® 710
Klon M1/70
Reaktivita Baboon,Cynomolgus monkey,Rhesus monkey,Chimpanzee,Human,Mouse
Hostitel Rat
Identifikační číslo genu 16409 / 3684
Izotyp IgG2b
Průtoková cytometrie Yes
Neutralizace Yes
Skladovací teplota Store Undiluted at 2 to 8 °C