Tisk…

Ethanolamine hydrochloride

Dodavatel: Merck

Synonyms: 2-Aminoethanol hydrochloride, 2-Hydroxyethylammonium Chloride, Monoethanolamine Hydrochloride, 2-Aminoethan-1-ol hydrochloride

M-Clarity™ quality level = MQ200

8.20063.0100 8.20063.0500
8.20063.0100EA 1010 CZK
8.20063.0100 8.20063.0500
Ethanolamine hydrochloride
Ethanolamine hydrochloride
Vzorec: C₂H₈ClNO
MW: 97.54
MDL Number: MFCD00012892
CAS číslo: 2002-24-6
EINECS: 217-900-6

Objednat

Výsledky specifikačního testu

Solubility (20 °C) soluble
Melting Point 80 - 84 °C
Molar Mass 97.54 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR