Tisk…

Glycerin for synthesis, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol

M-Clarity™ quality level = MQ200

8.18709.1000 8.18709.5000
8.18709.1000EA 1800 CZK
8.18709.1000 8.18709.5000
Glycerin for synthesis, Sigma-Aldrich®
Glycerin
Vzorec: HOCH₂CH(OH)CH₂OH
Bod varu: 290 °C (1013 hPa)
Hustota: 1,26 g/cm³ (20 °C)
Bod vzplanutí: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS číslo: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR