Tisk…

Nerolidol (cis and trans mixture) for synthesis, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol, 3-Hydroxy-3,7,11-trimethyl-1,6,10-dodecatriene, Nerolidol, 3,7,11-Trimethyldodeca-1,6,10-trien-3-ol

M-Clarity™ quality level = MQ200

8.18553.0100
8.18553.0100EA 2540 CZK
8.18553.0100
Nerolidol (cis and trans mixture) for synthesis, Sigma-Aldrich®
Nerolidol (cis and trans mixture)
Vzorec: C₁₅H₂₆O
Storage Temperature: Ambient
MDL Number: MFCD00008911
CAS číslo: 7212-44-4
EINECS: 230-597-5
UN: 3082
ADR: 9,III

Objednat

Výsledky specifikačního testu

Solubility 0,014 g/l (20 °C)
Melting Point -75 °C
Molar Mass 222.37 g/mol
Boiling Point 263 - 266 °C (1013 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 96 °C
Refractive Index 1.4893 (20 °C, 589 nm)
Explosion Limit 0.7 - 4.5 %(V)
Density 0.88 g/cm3 (20 °C)
Ignition Point 230 °C
Melting point -75 °C
Vapour pressure -1 hPa (20 °C)
Refractive index 1,4893 (20 °C, 589 nm)
Identity (IR) Passes test
Storage temperature +15...+25 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR