Tisk…

Potassium hydrogen monopersulphate for synthesis, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

8.18401.0100 8.18401.0500
8.18401.0100EA 837 CZK
8.18401.0100 8.18401.0500
Potassium hydrogen monopersulphate for synthesis, Sigma-Aldrich®
Potassium hydrogen monopersulphate
Vzorec: 2KHSO₅·KHSO₄·K₂SO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00040551
CAS číslo: 70693-62-8
EINECS: 274-778-7
UN: 3260
ADR: 8,III

Objednat

Výsledky specifikačního testu

Hill Formula H₃K₅O₁₈S₄
Chemical formula 2KHSO₅ * KHSO₄ * K₂SO₄
Molar Mass 614.76 g/mol
Density 1.1 - 1.2 g/cm³ (20 °C)
pH value 2 - 3 (10 g/l, H₂O, 20 °C)
Solubility 256 g/l (20 °C)
LD 50 oral LD50 Rat 2000 mg/kg
Storage class 8 B Non-combustible corrosive substances
WGK WGK 1 slightly water endangering
R Phrase R 8-34-37
S Phrase S 17-26-36/37/39-45


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR