Tisk…

Tetra-tert-butyl orthotitanate for synthesis, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ200

8.18320.0025
8.18320.0025EA 2810 CZK
8.18320.0025
Tetra-tert-butyl orthotitanate for synthesis, Sigma-Aldrich®
Tetra-tert-butyl orthotitanate
Vzorec: C₁₆H₃₆O₄Ti
Storage Temperature: Ambient
MDL Number: MFCD00040554
CAS číslo: 3087-39-6
EINECS: 221-412-9
UN: 1993
ADR: 3,III

Objednat

Výsledky specifikačního testu

Solubility (20 °C) (decomposition)
Molar Mass 340.32 g/mol
Boiling Point 160 - 165 °C (20 hPa)
Vapor Pressure 43 hPa (136 °C)
Refractive Index 1.4368 (20 °C, 589 nm)
Density 0.89 g/cm3 (20 °C)
Synonyms tert-Butyl titanate(IV), Orthotitanic acid tetra-tert-butyl ester, Titanium tetra-tert-butylate, Titanic acid tetra-tert-butyl ester
Hill Formula C16H36O4Ti
Chemical Formula Ti(OC(CH3)3)4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR