Tisk…

Dipentaerythritol for synthesis, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

8.14810.0025 8.14810.0100 8.14810.0500
8.14810.0025EA 1120 CZK
8.14810.0025 8.14810.0100 8.14810.0500
Dipentaerythritol for synthesis, Sigma-Aldrich®
Dipentaerythritol
Vzorec: C₁₀H₂₂O₇
MDL Number: MFCD00004691
CAS číslo: 126-58-9
EINECS: 204-794-1

Objednat

Výsledky specifikačního testu

Solubility 2 g/l (20 °C)
Melting Point 217 - 222 °C
Molar Mass 254.28 g/mol
Bulk Density 400 - 450 kg/m3
Vapor Pressure  - 0.00001 hPa (20 °C)
Density 1.365 g/cm3 (20 °C)
pH 5 (2 g/l, H2O, 20 °C)
Ignition Point 370 °C
Synonyms 2.2'[Oxybis(methylen)]bis[hydroxymethyl)]-1,3-propandiol
Hill Formula C10H22O7


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR