Tisk…

8-Hydroxyquinoline-5-sulphonic acid monohydrate for synthesis, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 8-Hydroxyquinoline-5-sulphonic acid hydrate

M-Clarity™ quality level = MQ200

8.14436.0100 8.14436.0500
8.14436.0100EA 3630 CZK
8.14436.0100 8.14436.0500
8-Hydroxyquinoline-5-sulphonic acid monohydrate for synthesis, Sigma-Aldrich®
8-Hydroxyquinoline-5-sulphonic acid monohydrate
Vzorec: C₉H₇NO₄S·1H₂O
MDL Number: MFCD00149482
CAS číslo: 283158-18-9
EINECS: 201-570-5

Objednat

Výsledky specifikačního testu

Solubility (20 °C) slightly soluble
Melting Point 322 °C
Molar Mass 243.24 g/mol
Hill Formula C9H7NO4S*H2O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR