Tisk…

Titanium(III) chloride 15% in hydrochloric acid 10% for synthesis, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

8.08307.0100 8.08307.0500 8.08307.1000
8.08307.0100EA 1530 CZK
8.08307.0100 8.08307.0500 8.08307.1000
Titanium(III) chloride 15% in hydrochloric acid 10% for synthesis, Sigma-Aldrich®
Titanium(III) chloride
Vzorec: Cl₃Ti
Hustota: 1,21 g/cm³ (20 °C)
MDL Number: MFCD00011266
CAS číslo: 7705-07-9
EINECS: 231-728-9
UN: 1789
ADR: 8,II

Objednat

Výsledky specifikačního testu

Assay (cerimetric TiCl₃) 14.5 - 15.5 %
Density 1.21 g/cm3 (20 °C)
pH  - 1 (H2O, 20 °C) (undiluted)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR