Tisk…

Polyethylenglykol 300 for synthesis, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: PEG 300

M-Clarity™ quality level = MQ200

8.07484.1000 8.07484.5000
8.07484.1000EA 1430 CZK
8.07484.1000 8.07484.5000
Polyethylenglykol 300 for synthesis, Sigma-Aldrich®
Polyethylenglykol 300
Vzorec: H(OCH₂CH₂)nOH
MW: 300
Bod tání: –15…–10 °C
Hustota: 1.13 g/cm³ (20 °C)
Bod vzplanutí: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS číslo: 25322-68-3
EINECS: 500-038-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR