Tisk…
Protein concentrator, Tube-O-CONCENTRATOR™
Dodavatel: G-Biosciences
786-625 786-626
786-625EA 11370 CZK
786-625 786-626
Protein concentrator, Tube-O-CONCENTRATOR™
Zkumavky Zkumavky dialyzační
Tube-O-CONCENTRATOR™ Mini for 20 to 250 μl volumes or Medi for 0,2 to 2,5 ml volumes.
  • Rapid concentration of protein solutions without protein precipitation
  • Combination of a unique tube format dialysis device and a water absorbing liquid polymer
  • Suitable for up to 250 µl or 2,5 ml protein solutions
  • Zero protein loss, even if protein precipitation occurs
Tube-O-CONCENTRATOR™ is a versatile concentration device that utilizes a novel, water absorbing, liquid polymer and our patented Tube-O-DIALYZER™ for the rapid concentration of dilute protein solutionsMore Product Information