Tisk…

Dihydrogenfosforečnan draselný 1 mol/l aqueous solution

Dodavatel: G Bioscienes

Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

786-488
786-488EA 4570 CZK
786-488
Dihydrogenfosforečnan draselný 1 mol/l aqueous solution
Dihydrogenfosforečnan draselný
Vzorec: KH₂PO₄
MW: 136.09
Bod varu: 100
Bod tání: 253
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS číslo: 7778-77-0
EINECS: 231-913-4
Merck Index: 13,07744

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR