Tisk…
Electrophoresis gel stain, Reversible Copper Stain™
Dodavatel: G Bioscienes
786-32CU 786-32DSCU
786-32CUEA 12810 CZK
786-32CU 786-32DSCU
Electrophoresis gel stain, Reversible Copper Stain™
Barviva Barviva pro elektroforézu
The Reversible Copper Stain™ is a single step stain for the rapid detection of proteins resolved on SDS-PAGE.
  • Based on the interaction of copper ions with polyacrylamide and proteins
  • Copper metal precipitate in the gel resulting in an opaque blue/green gel
  • Sensitivity of the Reversible Copper Stain™ is 0,1 to 0,5 ng BSA protein
Suitable for staining of protein gels prior to transfer or electroelution of proteins.
Upozornení: Not suitable for native gels or gels containing Tricine or Glycine.