Tisk…
Non-animal protein blocking Agent, NAP-BLOCKER™
Dodavatel: G Bioscienes
786-190 786-190P 786-190T
786-190EA 12380 CZK
786-190 786-190P 786-190T
Non-animal protein blocking Agent, NAP-BLOCKER™
Pufry Pufry pro elektroforézu Pufry blokovací
NAP-BLOCKER™ is an animal-free protein blocking agent recommended for improved assay sensitivity, minimal non specific binding and a high signal-to-background ratio. It is suitable for Western blots, dot blots and assay development.
  • Free from animal source blocking agents
  • Eliminates cross reactivity
  • Eliminates non specific binding