Tisk…

Hydroxid sodný ≥98% (by acidimetry), pellets, AR® ACS, Macron Fine Chemicals™

Dodavatel: Avantor Performance Materials
Nebezpečí

Synonyms: Caustic soda

7708-06EA 5630 CZK
7708-06 7708-20 7708-28
Hydroxid sodný ≥98% (by acidimetry), pellets, AR® ACS, Macron Fine Chemicals™
Hydroxid sodný

AR - The standard Macron Fine Chemicals™ grade of analytical reagents; suitable for laboratory and general use.

Vzorec: NaOH
MW: 40
Bod varu: 1390
Bod tání: 323
Hustota: 2.13
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS číslo: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8 II
REACH: 01-2119457892-27
Merck Index: 14,08627

Objednat

Výsledky specifikačního testu

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 98 %
Assay (NaOH) (by acidimetry) ≥98 %
Identification Passes Test
Calcium (Ca) ≤0.005 %
Chloride (Cl) ≤0.005 %
Copper (Cu) ≤0.001 %
Heavy Metals (as Ag) ≤0.001 %
Insoluble Matter ≤0.003 %
Iron (Fe) ≤0.0003 %
ACS - Magnesium (Mg) ≤0.002 %
Mercury (Hg) ≤0.1 ppm
Nickel (Ni) ≤0.0005 %
Nitrogen Compounds (as N) ≤0.0003 %
Phosphate (PO₄) ≤0.0002 %
Potassium (K) ≤0.02 %
Sodium Carbonate (Na₂CO₃) ≤0.4 %
ACS - Sulfate (SO₄) ≤0.003 %
Meets Reagent Specifications for testing USP/NF monographs
Appearance (white hygroscopic pellets)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR