Tisk…
Communicating tubes
Dodavatel: GLASWARENFABRIK KARL HECHT
41954010
765-0969EA 1840 CZK
765-0969
Communicating tubes
Materiály výukové Materiály pro výuku fyziky Aktivity pro výuku mechaniky
Five glass tubes of different shape and inner dimensions. Down to each other. When filling the liquid in the pipes, it is clear that the height of the stowage in the tubes. pipes are independent of the design of the pipes. Comes with foot.