Tisk…

Anti-CD24 Rat Monoclonal Antibody (violetFluor™ 450) [clone: M1/69]

Dodavatel: Tonbo Biosciences
75-0242-U025 75-0242-U100
75-0242-U025EA 2440 CZK
75-0242-U025 75-0242-U100
Anti-CD24 Rat Monoclonal Antibody (violetFluor™ 450) [clone: M1/69]
Protilátky
The M1/69 antibody reacts with mouse CD24, a cell surface molecule also known as Heat Stable Antigen (HSA). CD24 is a GPI-linked glycoprotein involved in cell adhesion and is a ligand for CD62P (P-selectin). It is expressed on erthrocytes, thymocytes, peripheral lymphocytes and myeloid lineage cells. The expression of CD24 is used to delineate stages of of lymphocyte development in the bone marrow.

Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: M1/69
Conjugation: violetFluor™ 450
Epitope:
Host: Rat
Isotype: Rat IgG2b, kappa
Reactivity: Mouse
Objednat

Produktová specifikace

Symbol antigenu Heat Stable Antigen, HSA
Název antigenu CD24, Heat Stable Antigen
Environmentally Preferable
Druh protilátky Primární
Klonovalita Monoclonal
Klon M1/69
Reaktivita Mouse
Hostitel Rat
Identifikační číslo genu 12484
Izotyp IgG2b kappa
Průtoková cytometrie Yes
Skladovací teplota 2-8°C protected from light

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR